Learn More
Cycloheximide solution, 100mg/mL in DMSO, 0.2 micron filtered, Thermo Scientific Chemicals
Cycloheximide, CAS # 66-81-9, is a compound produced by the bacterium Streptomyces griseus that shows an antifungal antibiotic activity with the ability to inhibit protein synthesis only in eukaryotic cells.
Supplier: Thermo Scientific Chemicals J66004XF
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cycloheximide | |
100mg/mL in DMSO, 0.2 micron filtered | |
66-81-9 | |
MFCD00082346 | |
CHX | |
[H][C@]1(C[C@@H](C)C[C@H](C)C1=O)[C@H](O)CC1CC(=O)NC(=O)C1 | |
281.35 | |
CHEBI:27641 | |
Colorless to Yellow | |
87°0°C (230°F) |
Cycloheximide solution | |
Liquid | |
C15H23NO4 | |
UN2810 | |
YPHMISFOHDHNIV-QTEFRXOUNA-N | |
4-[(2R)-2-[(1S,3S,5S)-3,5-dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidine-2,6-dione | |
6197 | |
281.35 | |
1 mL |
Chemical Identifiers
66-81-9 | |
281.35 | |
YPHMISFOHDHNIV-QTEFRXOUNA-N | |
6197 | |
4-[(2R)-2-[(1S,3S,5S)-3,5-dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidine-2,6-dione |
C15H23NO4 | |
MFCD00082346 | |
CHX | |
CHEBI:27641 | |
[H][C@]1(C[C@@H](C)C[C@H](C)C1=O)[C@H](O)CC1CC(=O)NC(=O)C1 |
Safety and Handling
GHS H Statement
H300-H360-H341-H227
Fatal if swallowed.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Combustible liquid.
P201-P202-P210-P264b-P270-P281-P301+P310-P308+P313-P330-P370+P378q-P501c
H227-H300-H341-H360D
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber : 200-636-0
RTECSNumber : MA4375000
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only