Please login to your online account to display your discounted pricing

Cyclohexene Sulfide (Tech.), ca 85%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  111220050

 View more versions of this product

Catalog No. AC111220050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Color Yellow
Formula Weight 114.2
Assay ≥80%
Physical Form Opalescent Liquid
Purity Grade Technical Grade
Quantity 5g
EINECS Number 206-008-2
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Flammable liquid and vapor.
Chemical Name or Material Cyclohexene sulfide
Assay Percent Range ca. 85%
CAS 286-28-2
Name Note tech., ca. 85%
MDL Number MFCD00005163
Molecular Formula C6H10S
Synonym 7-Thiabicyclo[4.1.0]heptane
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Warning

Specifications

Chemical Name or Material Cyclohexene sulfide
Name Note tech., ca. 85%
CAS 286-28-2
Purity Grade Technical Grade
Assay ≥80%
Assay Percent Range ca. 85%
Molecular Formula C6H10S
Formula Weight 114.2
MDL Number MFCD00005163
Synonym 7-Thiabicyclo[4.1.0]heptane
Accessories

GHS Signal Word: Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapor.

SDS & Safety Documents