Please login to your online account to display your discounted pricing

Cyclohexene Sulfide (Tech.), ca 85%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Cyclohexene sulfide
Name Note tech., ca. 85%
CAS 286-28-2
Purity Grade Technical Grade
Assay ≥80%
Assay Percent Range ca. 85%
Molecular Formula C6H10S
Formula Weight 114.2
MDL Number MFCD00005163
Synonym 7-Thiabicyclo[4.1.0]heptane
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC111220050

  SDS
thermo fisher scientific
111220050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC111220100

  SDS
thermo fisher scientific
111220100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Opalescent Liquid
Assay ≥80%
Formula Weight 114.2
Color Yellow
Purity Grade Technical Grade
Quantity 5g
EINECS Number 206-008-2
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Flammable liquid and vapor.
Chemical Name or Material Cyclohexene sulfide
Name Note tech., ca. 85%
Assay Percent Range ca. 85%
Molecular Formula C6H10S
MDL Number MFCD00005163
CAS 286-28-2
Synonym 7-Thiabicyclo[4.1.0]heptane
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapor.