Learn More
Cyclohexene, 99%, pure, stabilized, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 154845000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cyclohexene | |
-104.0°C | |
0.8100g/mL | |
−20°C | |
98.5% min. (GC) | |
C6H10 | |
C6H10 | |
05,63 | |
15,272 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol, miscible with all common organic solvents | |
C1CCC=CC1 | |
82.13 | |
0.66 mPa.s (22°C) | |
82.13 | |
Pure |
110-83-8 | |
Colorless | |
83.0°C | |
Authentic | |
Glass bottle | |
1.4450 to 1.4470 | |
500 mL | |
0.81 | |
tetrahydrobenzene, cyclohex-1-ene, benzene tetrahydride, 1,2,3,4-tetrahydrobenzene, benzenetetrahydride, hexanaphthylene, 1-cyclohexene, cykloheksen, benzene, tetrahydro, 3,4,5,6-tetrahydrobenzene | |
HGCIXCUEYOPUTN-UHFFFAOYSA-N | |
cyclohexene | |
8079 | |
CHEBI:36404 | |
99% | |
Liquid |
Chemical Identifiers
110-83-8 | |
tetrahydrobenzene, cyclohex-1-ene, benzene tetrahydride, 1,2,3,4-tetrahydrobenzene, benzenetetrahydride, hexanaphthylene, 1-cyclohexene, cykloheksen, benzene, tetrahydro, 3,4,5,6-tetrahydrobenzene | |
CHEBI:36404 | |
C1CCC=CC1 |
HGCIXCUEYOPUTN-UHFFFAOYSA-N | |
8079 | |
cyclohexene |
Safety and Handling
GHS H Statement
May be fatal if swallowed and enters airways.
Highly flammable liquid and vapor.
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi
GHS Signal Word: Danger
EINECSNumber : 203-807-8
RUO – Research Use Only