missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclohexene, 99%

Manufacturer:  Alfa Aesar™ A11359AE

 View more versions of this product

Catalog No. AAA11359AE



Description

Description

Applications
Cyclohexene is used as a solvent. It is employed as a precursor to caprolactam, adipic acid, maleic acid, dicyclohexyladipate and cyclohexeneoxide. It is used as an intermediate for the production of cyclohexanol and cyclohexeneoxide.

Solubility
Immiscible with cold water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Cyclohexene
Petrol-like
-104°C
110-83-8
MFCD00001539
906737
tetrahydrobenzene, cyclohex-1-ene, benzene tetrahydride, 1,2,3,4-tetrahydrobenzene, benzenetetrahydride, hexanaphthylene, 1-cyclohexene, cykloheksen, benzene, tetrahydro, 3,4,5,6-tetrahydrobenzene
C1CCC=CC1
82.146
CHEBI:36404
99%
−20°C (−4°F)
83°C to 85°C
100mL
C6H10
UN2256
14,2727
HGCIXCUEYOPUTN-UHFFFAOYSA-N
cyclohexene
8079
82.15

Chemical Identifiers

110-83-8
82.146
HGCIXCUEYOPUTN-UHFFFAOYSA-N
8079
cyclohexene
C6H10
MFCD00001539
tetrahydrobenzene, cyclohex-1-ene, benzene tetrahydride, 1,2,3,4-tetrahydrobenzene, benzenetetrahydride, hexanaphthylene, 1-cyclohexene, cykloheksen, benzene, tetrahydro, 3,4,5,6-tetrahydrobenzene
CHEBI:36404
C1CCC=CC1
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304-H302
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Harmful if swallowed.

GHS P Statement
P210-P280-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CYCLOHEXENE

RTECSNumber : GW2500000

TSCA : TSCA

SDS Documents