missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cyclohexene-1-boronic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H53247MD
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Cyclohexene-1-boronic acid | |
165°C | |
MFCD02179497 | |
cyclohex-1-en-1-ylboronic acid, 1-cyclohexenylboronic acid, 1-cyclohexen-1-yl-boronic acid, cyclohexenylboronic acid, cyclohexen-1-yl-boronic acid, 1-cyclohexen-1-ylboronic acid, cyclohexene-1-boronic acid, 1-boronocyclohex-1-ene, cyclohex-1-enylboronic acid, cyclohexen-1-yl boronic acid | |
OB(O)C1=CCCCC1 | |
125.96 | |
125.96 |
89490-05-1 | |
C6H11BO2 | |
250 mg | |
XZWQKJXJNKYMAP-UHFFFAOYSA-N | |
cyclohexen-1-ylboronic acid | |
3862902 | |
97% |
Chemical Identifiers
89490-05-1 | |
125.96 | |
XZWQKJXJNKYMAP-UHFFFAOYSA-N | |
3862902 | |
OB(O)C1=CCCCC1 |
C6H11BO2 | |
MFCD02179497 | |
cyclohex-1-en-1-ylboronic acid, 1-cyclohexenylboronic acid, 1-cyclohexen-1-yl-boronic acid, cyclohexenylboronic acid, cyclohexen-1-yl-boronic acid, 1-cyclohexen-1-ylboronic acid, cyclohexene-1-boronic acid, 1-boronocyclohex-1-ene, cyclohex-1-enylboronic acid, cyclohexen-1-yl boronic acid | |
cyclohexen-1-ylboronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Store at -20°C