Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclohexanone diethyl ketal, 98%

Chemical Identifiers

Chemical Name or Material Cyclohexanone diethyl ketal
CAS 1670-47-9
Molecular Formula C10H20O2
Formula Weight 172.27
MDL Number MFCD00037723
UN Number UN1993
Beilstein 1851801
Synonym 1,1-Diethoxycyclohexane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1416309

 
alfa aesar™
A1416309
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1416318

 
alfa aesar™
A1416318
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1416330

 
alfa aesar™
A1416330
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 172.27
Chemical Name or Material Cyclohexanone diethyl ketal
TSCA TSCA
Assay 98%
Molecular Formula C10H20O2
MDL Number MFCD00037723
CAS 1670-47-9
UN Number UN1993
Beilstein 1851801
Synonym 1,1-Diethoxycyclohexane
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Flash Point 58°C (136°F)
Odor Characteristic
Boiling Point 76° to 78°C (20mmHg)
Quantity 10g
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapour.