missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclohexanone (Certified), Fisher Chemical

$248.31 - $448.38

Chemical Identifiers

CAS 108-94-1
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00001625
InChI Key JHIVVAPYMSGYDF-UHFFFAOYSA-N
Synonym ketohexamethylene, pimelic ketone, sextone, cyclohexyl ketone, nadone, anone, anon, cyclohexanon, hytrol o, hexanon
PubChem CID 7967
ChEBI CHEBI:17854
IUPAC Name cyclohexanone
SMILES C1CCC(=O)CC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
O2109-1 SDS View Documents Thermo Scientific
O21091
1L Each for $248.31
Case of 6 Each for $1,489.86
O2109-4 SDS Accessories View Documents Thermo Scientific
O21094
4L Each for $448.38
Case of 4 Each for $1,793.52
Specifications

Chemical Identifiers

108-94-1
98.145
JHIVVAPYMSGYDF-UHFFFAOYSA-N
7967
cyclohexanone
C6H10O
MFCD00001625
ketohexamethylene, pimelic ketone, sextone, cyclohexyl ketone, nadone, anone, anon, cyclohexanon, hytrol o, hexanon
CHEBI:17854
C1CCC(=O)CC1

Specifications

108-94-1
≥98 %
MFCD00001625
ketohexamethylene, pimelic ketone, sextone, cyclohexyl ketone, nadone, anone, anon, cyclohexanon, hytrol o, hexanon
JHIVVAPYMSGYDF-UHFFFAOYSA-N
cyclohexanone
7967
98.14
≥98%
0.947g/cm³
Glass Bottle
4.5mbar at 20°C
Colorless
1L
0.05% max.
C6H10O
1915
2°C max.
C1CCC(=O)CC1
98.145
CHEBI:17854
Liquid
Certified
Pass Test
0.05% max.
155°C
-47°C
Cyclohexanone
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Flammable liquid and vapor. Irritating to skin. Risk of serious damage to eyes. Use only under a chemical fume hood. Wear personal protective equipment. Do not get in eyes, on skin, or on clothing. Avoid ingestion and inhalation. Keep away from open flames, hot surfaces and sources of ignition. Use only non-sparking tools. Use explosion-proof equipment. Take precautionary measures against static discharges. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required.

NFPA

Health:2
Flammability:2
Instability:0

DOTInformation : DOT Class 3, : Flammable Liquid