Please login to your online account to display your discounted pricing

Cyclohexanone (Certified), Fisher Chemical


Quantity 4L

Manufacturer: fisher scientific company  O21094

 View more versions of this product

Catalog No. O2109-4

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Boiling Point 155°C
Assay ≥98%
Formula Weight 98.14
Purity Grade Certified Grade
Melting Point −47°C
Quantity 4L
DOT Information DOT Class 3, : Flammable Liquid
Physical Form Liquid
Color Colorless
Molecular Formula C6H10O
Vapor Pressure 4.5mbar at 20°C
Density 0.947g/cm³
Residue after Evaporation 0.05% max.
Packaging Glass Bottle
Acidity 0.05% max.
Identification Pass Test
Assay Percent Range ≥98 %
Synonym Ketohexamethylene
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. Irritating to skin. Risk of serious damage to eyes. Use only under a chemical fume hood. Wear personal protective equipment. Do not get in eyes, on skin, or on clothing. Avoid ingestion and inhalation. Keep away from open flames, hot surfaces and sources of ignition. Use only non-sparking tools. Use explosion-proof equipment. Take precautionary measures against static discharges. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required.

NFPA

Health:2
Flammability:2
Instability:0
CAS 108-94-1
Chemical Name or Material Cyclohexanone
MDL Number MFCD00001625
UN Number 1915
Boiling Range 2°C max.
Cleanup Symbol Solvent
Health Hazard 2 DANGER!
ChemAlert Storage Symbol Red

Specifications

Chemical Name or Material Cyclohexanone
CAS 108-94-1
Purity Grade Certified Grade
Assay ≥98%
Assay Percent Range ≥98 %
Molecular Formula C6H10O
Formula Weight 98.14
MDL Number MFCD00001625
Synonym Ketohexamethylene
Accessories

Accessories for Catalog Number: O21094

Item Mfr. No. Item Description Price Per Unit Quantity  
03-439
Eagle™ Thermoplastics™ Safety Bottle Carriers
fisher scientific
B102
Safety bottle carriers are molded-in handle Each for $217.50
Accessories
 

Emergency Overview
Flammable liquid and vapor. Irritating to skin. Risk of serious damage to eyes. Use only under a chemical fume hood. Wear personal protective equipment. Do not get in eyes, on skin, or on clothing. Avoid ingestion and inhalation. Keep away from open flames, hot surfaces and sources of ignition. Use only non-sparking tools. Use explosion-proof equipment. Take precautionary measures against static discharges. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required.

NFPA

Health:2
Flammability:2
Instability:0

DANGER!

 

SDS & Safety Documents