Please login to your online account to display your discounted pricing

Cyclohexanone (Certified), Fisher Chemical


Quantity 1L

Manufacturer: fisher scientific company  O21091

 View more versions of this product

Catalog No. O2109-1

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98%
Purity Grade Certified Grade
Boiling Point 155°C
Melting Point −47°C
Formula Weight 98.14
Quantity 1L
DOT Information DOT Class 3, : Flammable Liquid
Vapor Pressure 4.5mbar at 20°C
Physical Form Liquid
Color Colorless
Molecular Formula C6H10O
Density 0.947g/cm³
Residue after Evaporation 0.05% max.
CAS 108-94-1
ChemAlert Storage Symbol Red
Synonym Ketohexamethylene
Health Hazard 2 DANGER!
Acidity 0.05% max.
Chemical Name or Material Cyclohexanone
Packaging Glass Bottle
MDL Number MFCD00001625
UN Number 1915
Identification Pass Test
Boiling Range 2°C max.
Cleanup Symbol Solvent
Assay Percent Range ≥98 %
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. Irritating to skin. Risk of serious damage to eyes. Use only under a chemical fume hood. Wear personal protective equipment. Do not get in eyes, on skin, or on clothing. Avoid ingestion and inhalation. Keep away from open flames, hot surfaces and sources of ignition. Use only non-sparking tools. Use explosion-proof equipment. Take precautionary measures against static discharges. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required.

NFPA

Health:2
Flammability:2
Instability:0

Specifications

Chemical Name or Material Cyclohexanone
CAS 108-94-1
Purity Grade Certified Grade
Assay ≥98%
Assay Percent Range ≥98 %
Molecular Formula C6H10O
Formula Weight 98.14
MDL Number MFCD00001625
Synonym Ketohexamethylene
Accessories
 

Emergency Overview
Flammable liquid and vapor. Irritating to skin. Risk of serious damage to eyes. Use only under a chemical fume hood. Wear personal protective equipment. Do not get in eyes, on skin, or on clothing. Avoid ingestion and inhalation. Keep away from open flames, hot surfaces and sources of ignition. Use only non-sparking tools. Use explosion-proof equipment. Take precautionary measures against static discharges. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required.

NFPA

Health:2
Flammability:2
Instability:0

DANGER!

 

SDS & Safety Documents