Please login to your online account to display your discounted pricing

Cyclohexanone, 99.8%, extra pure, ACROS Organics™


Quantity 25L
Packaging Metal drum

Manufacturer: thermo fisher scientific  AC111190251

 View more versions of this product

Catalog No. AC111190251

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade Extra Pure
Physical Form Liquid
Assay ≥99.75%
Formula Weight 98.14
Quantity 25L
Color Clear
Beilstein 07, 8
Fieser 01,178
Merck Index 15, 2719
Density 0.9470g/mL
Chemical Name or Material Cyclohexanone
Refractive Index 1.4450 to 1.4520 (20°C, 589nm)
Water 0.2% max. (K.F.)
Linear Formula C6H10(=O)
CAS 108-94-1
Infrared Spectrum Authentic
Packaging Metal drum
Assay Percent Range 99.8%
Molecular Formula C6H10O
MDL Number MFCD00001625
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS P Statement
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye damage.
Causes skin irritation.

Specifications

Chemical Name or Material Cyclohexanone
CAS 108-94-1
Purity Grade Extra Pure
Assay ≥99.75%
Assay Percent Range 99.8%
Molecular Formula C6H10O
Linear Formula C6H10(=O)
Formula Weight 98.14
MDL Number MFCD00001625
Accessories

Danger

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye damage.
Causes skin irritation.

SDS & Safety Documents