missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclohexanol, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 147680100

Catalog No. AC147680100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Cyclohexanol
0.9600g/mL
67°C
98%
C6H12O
C6H11OH
06,5
15,2718
Solubility in water: 3.6g/100mL (20°C). Other solubilities: soluble in ethanol, ethyle acetate and, aromatic hydrocarbons
C1CCC(CC1)O
100.16
CHEBI:18099
98%
108-93-0
161.0°C
Authentic
Plastic drum
1.4630 to 1.4670
10L
0.96
cyclohexyl alcohol, hexahydrophenol, hydrophenol, hydroxycyclohexane, hexalin, 1-cyclohexanol, hydralin, adronal, naxol, adronol
HPXRVTGHNJAIIH-UHFFFAOYSA-N
cyclohexanol
7966
100.16
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful if inhaled.
Harmful to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protec

GHS Signal Word: Warning

EINECSNumber : 203-630-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only