Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclohexano-15-crown-5, cis + trans, 97%


Quantity 1g

Manufacturer: alfa aesar™  L1696403

 View more versions of this product

Catalog No. AAL1696403

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 274.36
Chemical Name or Material Cyclohexano-15-crown-5
CAS 17454-48-7
MDL Number MFCD00054545
Health Hazard 1 Warning
Synonym 2,3-Cyclohexano-1,4,7,10,13-pentaoxacyclopentadecane
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Assay 97%
Molecular Formula C14H26O5
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Quantity 1g
Name Note Cis + Trans
Beilstein 1682984
Boiling Point 180°C (0.4mmHg)
Flash Point >100°C (212°F)

Specifications

Chemical Name or Material Cyclohexano-15-crown-5
CAS 17454-48-7
Name Note Cis + Trans
Molecular Formula C14H26O5
Formula Weight 274.36
MDL Number MFCD00054545
Beilstein 1682984
Synonym 2,3-Cyclohexano-1,4,7,10,13-pentaoxacyclopentadecane
Accessories

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents