Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclohexano-15-crown-5, cis + trans, 97%

$93.27 - $93.27

Chemical Identifiers

Chemical Name or Material Cyclohexano-15-crown-5
CAS 17454-48-7
Name Note Cis + Trans
Molecular Formula C14H26O5
Formula Weight 274.36
MDL Number MFCD00054545
Beilstein 1682984
Synonym 2,3-Cyclohexano-1,4,7,10,13-pentaoxacyclopentadecane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1696403

 
alfa aesar™
L1696403
1g Each for $93.27
Description & Specifications

Specifications

Formula Weight 274.36
Chemical Name or Material Cyclohexano-15-crown-5
MDL Number MFCD00054545
CAS 17454-48-7
Assay 97%
Molecular Formula C14H26O5
Synonym 2,3-Cyclohexano-1,4,7,10,13-pentaoxacyclopentadecane
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Name Note Cis + Trans
Beilstein 1682984
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Flash Point >100°C (212°F)
Boiling Point 180°C (0.4mmHg)
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.