Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclohexanethiol, 99%

Chemical Identifiers

Chemical Name or Material Cyclohexanethiol
CAS 1569-69-3
Molecular Formula C6H12S
Formula Weight 116.23
MDL Number MFCD00001455
UN Number UN3054
Beilstein 1236342
Synonym Cyclohexyl mercaptan
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2165630

 
alfa aesar™
B2165630
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 116.23
Chemical Name or Material Cyclohexanethiol
TSCA TSCA
Assay 99%
Molecular Formula C6H12S
MDL Number MFCD00001455
CAS 1569-69-3
UN Number UN3054
Beilstein 1236342
Synonym Cyclohexyl mercaptan
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H226-H332-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapour.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Boiling Point 158° to 160°C
Melting Point −30°C
Quantity 250g
Flash Point 43°C (109°F)
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CYCLOHEXYL MERCAPTAN
RTECS Number GV7525000
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H226-H332-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapour.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.