Please login to your online account to display your discounted pricing

Cyclohexanepropionic acid, 99%, ACROS Organics™


Quantity 50g

Manufacturer: fisher scientific acros  171450500

 View more versions of this product

Catalog No. AC171450500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Health Hazard 3 GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Health Hazard 2 GHS H Statement
Causes serious eye irritation. Harmful if swallowed. Causes skin irritation.
Physical Form Liquid
Assay ≥98.5%
Formula Weight 156.22
Quantity 50g
Color Clear colorless to slightly yellow
Beilstein 09, 82
TSCA TSCA
EINECS Number 211-861-9
Refractive Index 1.4630 to 1.4650 (20°C, 589nm)
Infrared Spectrum Authentic
Health Hazard 1 Warning
CAS 701-97-3
Name Note 99%
Linear Formula C6H11CH2CH2CO2H
Chemical Name or Material Cyclohexanepropionic acid
Molecular Formula C9H16O2
MDL Number MFCD00001527
Assay Percent Range 99%
Storage Information Product may darken during storage
Packaging Glass bottle

Specifications

Chemical Name or Material Cyclohexanepropionic acid
Name Note 99%
CAS 701-97-3
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H16O2
Linear Formula C6H11CH2CH2CO2H
Formula Weight 156.22
MDL Number MFCD00001527
Accessories

Warning

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
Causes serious eye irritation. Harmful if swallowed. Causes skin irritation.

SDS & Safety Documents