Learn More
Cyclohexanemethanol, 99%
CAS: 100-49-2 | C7H14O | 114.188 g/mol
Supplier: Thermo Scientific Chemicals L0653822
Description
Cyclohexanemethanol used as solvents, fuels . It is also used in pharmaceuticals, plasticizers, surfactants, lubricants, ore floatation agents, hydraulic fluids, and detergents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCyclohexanemethanol used as solvents, fuels . It is also used in pharmaceuticals, plasticizers, surfactants, lubricants, ore floatation agents, hydraulic fluids, and detergents.
Solubility
Soluble in Ether, Alcohols.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents, Acid chlorides, Acid anhydrides.
Specifications
| Cyclohexanemethanol | |
| 0.926 | |
| 71°C (159°F) | |
| 1.464 | |
| 100 g | |
| 14,2731 | |
| Soluble in Ether,Alcohols. | |
| C1CCC(CC1)CO | |
| 114.188 | |
| 114.19 |
| 100-49-2 | |
| 180°C to 181°C | |
| C7H14O | |
| MFCD00001510 | |
| 773712 | |
| cyclohexanemethanol, cyclohexylcarbinol, hexahydrobenzyl alcohol, cyclohexanecarbinol, methanol, cyclohexyl, cyclohexylmethyl alcohol, benzyl alcohol, hexahydro, usaf do-49, hydroxymethylcyclohexane, hydroxymethyl cyclohexane | |
| VSSAZBXXNIABDN-UHFFFAOYSA-N | |
| cyclohexylmethanol | |
| 7507 | |
| 99% |
Chemical Identifiers
| 100-49-2 | |
| 114.188 | |
| VSSAZBXXNIABDN-UHFFFAOYSA-N | |
| 7507 | |
| C1CCC(CC1)CO |
| C7H14O | |
| MFCD00001510 | |
| cyclohexanemethanol, cyclohexylcarbinol, hexahydrobenzyl alcohol, cyclohexanecarbinol, methanol, cyclohexyl, cyclohexylmethyl alcohol, benzyl alcohol, hexahydro, usaf do-49, hydroxymethylcyclohexane, hydroxymethyl cyclohexane | |
| cyclohexylmethanol |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 202-857-8
RTECSNumber : GV5075000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only