missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclohexanecarboxylic acid, 98%

Manufacturer:  Alfa Aesar™ A146930B

 View more versions of this product

Catalog No. AAA146930B



Description

Description

Applications
Cyclohexanecarboxylic acid can be used to determine complex binding constants of the three native cyclodextrins with seven cyclohexane derivatives.

Solubility
Soluble in water 0.201 g/100 mL (15°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents and strong bases.
Specifications

Specifications

Cyclohexanecarboxylic acid
231°C to 233°C
1000g
C7H12O2
970529
hexahydrobenzoic acid, carboxycyclohexane, cyclohexanoic acid, cyclohexylcarboxylic acid, cyclohexylmethanoic acid, cyclohexylformic acid, benzoic acid, hexahydro, cyclohexancarbonsaeure, cyclohexane-1-carboxylate, cyclohexanecarboxylicacid
C1CCC(CC1)C(=O)O
128.171
CHEBI:36096
98%
118°C (244°F)
29°C to 32°C
98-89-5
MFCD00001461
14,2724
NZNMSOFKMUBTKW-UHFFFAOYSA-N
cyclohexanecarboxylic acid
7413
128.17

Chemical Identifiers

98-89-5
128.171
NZNMSOFKMUBTKW-UHFFFAOYSA-N
7413
cyclohexanecarboxylic acid
C7H12O2
MFCD00001461
hexahydrobenzoic acid, carboxycyclohexane, cyclohexanoic acid, cyclohexylcarboxylic acid, cyclohexylmethanoic acid, cyclohexylformic acid, benzoic acid, hexahydro, cyclohexancarbonsaeure, cyclohexane-1-carboxylate, cyclohexanecarboxylicacid
CHEBI:36096
C1CCC(CC1)C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : GU8370000

TSCA : TSCA

Documents