Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclohexanecarboxaldehyde, 97%

$79.41 - $79.41

Chemical Identifiers

Chemical Name or Material Cyclohexanecarboxaldehyde
CAS 2043-61-0
Molecular Formula C7H12O
Formula Weight 112.17
MDL Number MFCD00001457
UN Number UN1989
Beilstein 878306
Synonym Hexahydrobenzaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2383714

  SDS
alfa aesar™
B2383714
25g Each for $79.41
Description & Specifications

Specifications

Formula Weight 112.17
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALDEHYDES, N.O.S.
Assay 97%
CAS 2043-61-0
UN Number UN1989
Molecular Formula C7H12O
Synonym Hexahydrobenzaldehyde
Health Hazard 1 Warning
Boiling Point 161° to 163°C
Melting Point 34° to 35°C
Chemical Name or Material Cyclohexanecarboxaldehyde
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flash Point 40°C (105°F)
MDL Number MFCD00001457
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 878306
Quantity 25g

Applications
Cyclohexanecarboxaldehyde is used for the synthesis of poly(vinyl cyclohexanal). It was used as model substrate in the synthesis of propargylamines using Fe3O4 nanoparticles as catalyst. It was employed in an aldol reaction with ethyl diazoacetate catalyzed by basic, nanoparticulate magnesium oxide leading to α-diazo-β-hydroxyesters.

Solubility
Soluble in chloroform, ether, alcohol. Slightly soluble in water.

Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.