Please login to your online account to display your discounted pricing

Cyclohexane (Reagent), Fisher Chemical


Quantity 4L
Packaging Glass Bottle

Manufacturer: fisher scientific company  O20934

 View more versions of this product

Catalog No. O2093-4

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Reagent Grade
Boiling Point 81°C
Melting Point 6.5°C
Formula Weight 84.16
Quantity 4L
DOT Information DOT Class 3, : Flammable Liquid
Vapor Pressure 104mbar at 20°C
Viscosity 0.94 mPa/s at 20°C
Physical Form Liquid
Color Colorless
Molecular Formula C6H12
Density 0.770g/cm³
Identification Pass Test
Additional Information Vapor Pressure: 104mbar at 20°C
Packaging Glass Bottle
MDL Number MFCD00003814
Boiling Range 2.0°C max., 78.7° to 82.7°C
Synonym Hexahydrobenzene
Health Hazard 2 DANGER!
Chemical Name or Material Cyclohexane
Cleanup Symbol Solvent
Linear Formula CH2(CH2)4CH
ChemAlert Storage Symbol Red
Health Hazard 3 Emergency Overview
Extremely flammable liquid and vapor. Inhalation may cause central nervous system effects. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:1
Flammability:3
Instability:0
CAS 110-82-7

Specifications

Chemical Name or Material Cyclohexane
CAS 110-82-7
Purity Grade Reagent Grade
Molecular Formula C6H12
Linear Formula CH2(CH2)4CH
Formula Weight 84.16
MDL Number MFCD00003814
Synonym Hexahydrobenzene
Accessories

Accessories for Catalog Number: O20934

Item Mfr. No. Item Description Price Per Unit Quantity  
Accessories
 

Emergency Overview
Extremely flammable liquid and vapor. Inhalation may cause central nervous system effects. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:1
Flammability:3
Instability:0

DANGER!

 

SDS & Safety Documents