Please login to your online account to display your discounted pricing

Cyclohexane (Reagent), Fisher Chemical


Quantity 20L
Packaging Steel Pail

Manufacturer: fisher scientific company  O209320

 View more versions of this product

Catalog No. O2093-20

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Reagent Grade
Boiling Point 81°C
Melting Point 6.5°C
Formula Weight 84.16
Quantity 20L
DOT Information DOT Class 3, : Flammable Liquid
Vapor Pressure 104mbar at 20°C
Viscosity 0.94 mPa/s at 20°C
Physical Form Liquid
Color Colorless
Density 0.770g/cm³
Molecular Formula C6H12
Additional Information Vapor Pressure: 104mbar at 20°C
Boiling Range 2.0°C max., 78.7° to 82.7°C
Packaging Steel Pail
Cleanup Symbol Solvent
MDL Number MFCD00003814
ChemAlert Storage Symbol Red
Synonym Hexahydrobenzene
Health Hazard 3 Emergency Overview
Extremely flammable liquid and vapor. Inhalation may cause central nervous system effects. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:1
Flammability:3
Instability:0
Identification Pass Test
Health Hazard 2 DANGER!
Chemical Name or Material Cyclohexane
Linear Formula CH2(CH2)4CH
CAS 110-82-7

Specifications

Chemical Name or Material Cyclohexane
CAS 110-82-7
Purity Grade Reagent Grade
Molecular Formula C6H12
Linear Formula CH2(CH2)4CH
Formula Weight 84.16
MDL Number MFCD00003814
Synonym Hexahydrobenzene
Accessories
 

Emergency Overview
Extremely flammable liquid and vapor. Inhalation may cause central nervous system effects. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:1
Flammability:3
Instability:0

DANGER!

 

SDS & Safety Documents