Please login to your online account to display your discounted pricing

Cyclohexane (Reagent), Fisher Chemical

$411.00 - $1,828.85

Chemical Identifiers

Chemical Name or Material Cyclohexane
CAS 110-82-7
Purity Grade Reagent Grade
Molecular Formula C6H12
Linear Formula CH2(CH2)4CH
Formula Weight 84.16
MDL Number MFCD00003814
Synonym Hexahydrobenzene
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

O2093-20

  SDS
fisher scientific company
O209320
Steel Pail 20L Each for $1,828.85

O2093-4

  Accessories SDS
fisher scientific company
O20934
Glass Bottle 4L Each for $411.00
Case of 4 Each for $1,644.00
Description & Specifications

Specifications

Purity Grade Reagent Grade
Formula Weight 84.16
Melting Point 6.5°C
Boiling Point 81°C
Quantity 20L
DOT Information DOT Class 3, : Flammable Liquid
Physical Form Liquid
Vapor Pressure 104mbar at 20°C
Viscosity 0.94 mPa/s at 20°C
Density 0.770g/cm³
Color Colorless
Molecular Formula C6H12
Boiling Range 2.0°C max., 78.7° to 82.7°C
Additional Information Vapor Pressure: 104mbar at 20°C
Health Hazard 3 Emergency Overview
Extremely flammable liquid and vapor. Inhalation may cause central nervous system effects. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:1
Flammability:3
Instability:0
Cleanup Symbol Solvent
ChemAlert Storage Symbol Red
MDL Number MFCD00003814
Packaging Steel Pail
Synonym Hexahydrobenzene
Identification Pass Test
Health Hazard 2 DANGER!
Linear Formula CH2(CH2)4CH
Chemical Name or Material Cyclohexane
CAS 110-82-7

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling
 

Emergency Overview
Extremely flammable liquid and vapor. Inhalation may cause central nervous system effects. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:1
Flammability:3
Instability:0

DANGER!