missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclohexane, HPLC Grade, 99% min.; Alfa Aesar™

Manufacturer:  Alfa Aesar 22919K7

 View more versions of this product

Catalog No. AA22919K7



Specifications

Specifications

Cyclohexane
Colorless liquid
C6H12
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
C1CCCCC1
84.162
CHEBI:29005
Liquid
0.779g/cm3 at 20°C
Pungent
Filtered through 0.2μ filters.
4L
110-82-7
99% min.
MFCD00003814
XDTMQSROBMDMFD-UHFFFAOYSA-N
cyclohexane
8078
84.16
HPLC
Maximum level of impurities: Evaporation residue 3ppm, Chlorinated pesticides 5ppt, H2O 0.02%.
104 hPa (78mm Hg) at 20°C
Colorless

Chemical Identifiers

110-82-7
84.162
XDTMQSROBMDMFD-UHFFFAOYSA-N
8078
cyclohexane
C6H12
MFCD00003814
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
CHEBI:29005
C1CCCCC1
Safety and Handling

Safety and Handling

Keep container in a well ventilated place. Keep away from sources of ignition - No Smoking. Avoid contact with eyes. Take precautionary measures against static discharges. This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets If swallowed, do not induce vomiting: seek medical advice immediately & show this container or label.

H225-H304-H400-H410-H315-H336: Highly flammable liquid and vapour. May be fatal if swallowed and enters airways. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Causes skin irritation. May cause drowsiness or dizziness.

DOTInformation : DOT Class: 3; Packing Group: II

EINECSNumber : 203-806-2

SDS Documents