Please login to your online account to display your discounted pricing

Cyclohexane, HPLC Grade, 99%, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Cyclohexane
CAS 110-82-7
Purity Grade HPLC Grade
Molecular Formula C6H12
Formula Weight 84.16
MDL Number MFCD00003814
UN Number UN1145
Beilstein 1900225
Merck Index 14,2723
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA22919M1

 
alfa aesar
22919M1
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade HPLC Grade
Formula Weight 84.16
Quantity 4x1L
Chemical Name or Material Cyclohexane
TSCA TSCA
Odor Pungent
Refractive Index 1.4260
Flash Point −18°C (−1°F)
Boiling Point 80° to 81°C
Physical Form Liquid
Color Colorless
Melting Point 6.5°C
Assay ≥99%
Molecular Formula C6H12
MDL Number MFCD00003814
CAS 110-82-7
UN Number UN1145
Beilstein 1900225
Merck Index 14,2723
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H304-H315-H336;
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Density 0.779
Health Hazard 3 GHS P Statement
P210-P261-P301+P310-P303+P361+P353-P405-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapours/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CYCLOHEXANE
RTECS Number GU6300000
Safety and Handling

Danger

GHS P Statement
P210-P261-P301+P310-P303+P361+P353-P405-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapours/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H304-H315-H336;
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.