Please login to your online account to display your discounted pricing

Cyclohexane, For Spectroscopy Uvasol™, EMD Millipore

$166.23 - $594.09

Chemical Identifiers

Chemical Name or Material Cyclohexane
CAS 110-82-7
Molecular Formula C6H12
Formula Weight 84.16
UN Number 1145
Synonym Hexahydrobenzene, Hexamethylene, Naphthene
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1028222500

 
emd millipore corporation
1028222500
Glass Bottle 2.5L Each for $594.09

M1028220500

 
emd millipore corporation
1028220500
Glass Bottle 500mL Each for $166.23
Description & Specifications

Specifications

Melting Point 6°C
Formula Weight 84.16
Boiling Point 81°C
Quantity 2.5L
Refractive Index 1.42
Flash Point −18°C (−0.4°F)
Chemical Name or Material Cyclohexane
Physical Form Liquid
Health Hazard 3 H315: Causes skin irritation.
UN Number 1145
Storage Information 5°C to 30°C (41°F to 86°F); Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition.
Ignition Point 260°C (500°F)
Synonym Hexahydrobenzene, Hexamethylene, Naphthene
Health Hazard 1 H225: Highly flammable liquid and vapour.
Solubility Information Soluble in water: 55mg/L at 20°C
Health Hazard 2 H304: May be fatal if swallowed and enters airways.
Molecular Formula C6H12
Color Colorless
CAS 110-82-7
Description The Uvasol™ solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The Uvasol™ solvents offer best UV transmittance.
Vapor Pressure 103 hPa (20°C)
Odor Charactristic
Packaging Glass Bottle
Density 0.78g/cm3 at 20°C
Safety and Handling

H225: Highly flammable liquid and vapour.

H315: Causes skin irritation.

H304: May be fatal if swallowed and enters airways.