missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cyclohexane, For Spectroscopy Uvasol™, MilliporeSigma™
Supplier: Technidata 1028220500
Specifications
Cyclohexane | |
110-82-7 | |
0.78g/cm3 at 20°C | |
500 mL | |
260°C (500°F) | |
Glass Bottle | |
1.42 | |
MFCD00003814 | |
1145 | |
Soluble in water: 55mg/L at 20°C | |
C1CCCCC1 | |
84.16 | |
CHEBI:29005 |
6°C | |
Colorless | |
81°C | |
−18°C (−0.4°F) | |
Charactristic | |
Liquid | |
C6H12 | |
103 hPa (20°C) | |
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane | |
XDTMQSROBMDMFD-UHFFFAOYSA-N | |
cyclohexane | |
8078 | |
84.16 |
Chemical Identifiers
110-82-7 | |
84.16 | |
XDTMQSROBMDMFD-UHFFFAOYSA-N | |
8078 | |
cyclohexane |
C6H12 | |
MFCD00003814 | |
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane | |
CHEBI:29005 | |
C1CCCCC1 |
Safety and Handling
H304: May be fatal if swallowed and enters airways.
H315: Causes skin irritation.
H225: Highly flammable liquid and vapour.
Recommended Storage : 5°C to 30°C (41°F to 86°F); Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition.