missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclohexane, For gas chromatography MS SupraSolv™, EMD Millipore

For gas chromatography MS SupraSolv™

$222.09 - $453.94

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Formula Weight 84.16
UN Number 1145
Synonym Hexahydrobenzene, Hexamethylene, Naphthene
Chemical Name or Material Cyclohexane
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
M1006671000 Emd Millipore Corporation
1006671000
Glass Bottle 1L Each for $222.09
M1006672500 Emd Millipore Corporation
1006672500
Glass Bottle 2.5L Each for $453.94
Specifications Description & Specifications

Chemical Identifiers

110-82-7
84.16
Hexahydrobenzene, Hexamethylene, Naphthene
C6H12
1145
Cyclohexane

Specifications

110-82-7
C6H12
1145
Soluble in water: 55mg/L at 20°C
0.78g/cm3 at 20°C
Charactristic
1.42
103 hPa (20°C)
81°C
6°C
Cyclohexane
Liquid
84.16
Hexahydrobenzene, Hexamethylene, Naphthene
SupraSolv™ MS is dedicated for use in gas chromatography coupled with mass spectrometric detection. SupraSolv™ solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results.
260°C (500°F)
Glass Bottle
5°C to 30°C (41°F to 86°F); Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition.
−18°C (−0.4°F)
Colorless
1L

Safety and Handling

H304: May be fatal if swallowed and enters airways.

H315: Causes skin irritation.

H225: Highly flammable liquid and vapour.