missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclohexane, EMD Millipore

For environmental analysis

Manufacturer:  Millipore Sigma CX22866

 View more versions of this product

Catalog No. MCX22866



Specifications

Specifications

Cyclohexane
C6H12
UN 1145
XDTMQSROBMDMFD-UHFFFAOYSA-N
cyclohexane
8078
84.18
≥99.0%
3.5
Pungent, fruity
Insoluble in water
Environmental analysis
Liquid
Colorless
1L
110-82-7
CH2(CH2)4CH2
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
C1CCCCC1
84.162
CHEBI:29005
Liquid
0.78g/cm3 at 20°C
1.2 to 8.3%(V)
Glass Bottle
103 hPa (20°C)
-20.15°C (-4.3°F)
80.6°C
6.7°C

Chemical Identifiers

110-82-7
84.162
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
CHEBI:29005
C1CCCCC1
C6H12
XDTMQSROBMDMFD-UHFFFAOYSA-N
8078
cyclohexane
Safety and Handling

Safety and Handling

DOTInformation : DOT Class: 3, Packaging Group: II

Recommended Storage : Store at room temperature.

Documents