Please login to your online account to display your discounted pricing

Cyclohexane-d12, for NMR, 99.5% atom D, ACROS Organics™


Quantity 25g

Manufacturer: thermo fisher scientific  AC166280250

 View more versions of this product

Catalog No. AC166280250

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 96.26
Physical Form Liquid
Quantity 25g
Color Clear colorless
Purity Grade For NMR
Density 0.8900g/mL
MDL Number MFCD00044212
Linear Formula C6D12
Molecular Formula C6D12
Chemical Name or Material Cyclohexane-d12
Name Note 99.5 atom%D
CAS 1735-17-7
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Deuterium (D) 99.4% min.
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
May cause drowsiness or dizziness.
May be fatal if swallowed and enters airways.
Highly flammable liquid and vapor.

Specifications

Chemical Name or Material Cyclohexane-d12
Name Note 99.5 atom%D
CAS 1735-17-7
Purity Grade For NMR
Molecular Formula C6D12
Linear Formula C6D12
Formula Weight 96.26
MDL Number MFCD00044212
Accessories

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
May cause drowsiness or dizziness.
May be fatal if swallowed and enters airways.
Highly flammable liquid and vapor.

SDS & Safety Documents