missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclohexane-d12, for NMR, 99.5% atom D, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 166280050

 View more versions of this product

Catalog No. AC166280050



Description

Description

For NMR
Specifications

Specifications

Cyclohexane-d12
99.4% min.
Glass bottle
0.89
Colorless
99.5 atom%D
C6H12
MFCD00044212
XDTMQSROBMDMFD-LBTWDOQPSA-N
1,1,2,2,3,3,4,4,5,5,6,6-dodecadeuteriocyclohexane
123129
Liquid
0.8900g/mL
−18°C
1.421
78.0°C
5g
1735-17-7
C6D12
2 h?? cyclohexane
C1CCCCC1
96.235
96.26
NMR

Chemical Identifiers

1735-17-7
96.235
XDTMQSROBMDMFD-LBTWDOQPSA-N
123129
C1CCCCC1
C6H12
MFCD00044212
2 h?? cyclohexane
1,1,2,2,3,3,4,4,5,5,6,6-dodecadeuteriocyclohexane
Safety and Handling

Safety and Handling

GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
May cause drowsiness or dizziness.
May be fatal if swallowed and enters airways.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents