Please login to your online account to display your discounted pricing

Cyclohexane-D12, deuteration degree min. 99.5%, for NMR spectroscopy, MagniSolv™, EMD Millipore™


For NMR Spectroscopy

Packaging Glass ampoule

Manufacturer: emd millipore corporation  8.15024.0005

 View more versions of this product

Catalog No. M8150240005

  • / Each
 

Description & Specifications

Chemical Identifiers

Odor Characterstic
Packaging Glass ampoule
Ignition Point 260°C
Vapor Pressure 103 hPa (20°C)
Quantity 10 x 0.5mL
Color Colorless
Chemical Name or Material Cyclohexane-D12
CAS 1735-17-7
Physical Form Liquid
Assay Percent Range 99.5% min.
Molecular Formula C6D12 (Hill)
Formula Weight 96.23 g/mol

Specifications

Chemical Name or Material Cyclohexane-D12
CAS 1735-17-7
Physical Form Liquid
Assay Percent Range 99.5% min.
Molecular Formula C6D12 (Hill)
Formula Weight 96.23 g/mol