Please login to your online account to display your discounted pricing

Cyclohexane-D12, deuteration degree min. 99.5%, for NMR spectroscopy, MagniSolv™, EMD Millipore™

For NMR Spectroscopy

$209.79 - $316.60

Chemical Identifiers

Chemical Name or Material Cyclohexane-D12
CAS 1735-17-7
Physical Form Liquid
Assay Percent Range 99.5% min.
Molecular Formula C6D12 (Hill)
Formula Weight 96.23
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M8150240005

 
emd millipore corporation
8150240005
Glass ampoule 10 x 0.5mL Each for $251.41

M8150240006

 
emd millipore corporation
8150240006
Glass ampoule 5mL Each for $209.79

M8150240009

 
emd millipore corporation
8150240009
Glass ampoule 10 x 0.75mL Each for $316.60
Description & Specifications

Specifications

Formula Weight 96.23
Quantity 10 x 0.5mL
Color Colorless
Physical Form Liquid
Assay Percent Range 99.5% min.
Odor Characterstic
Packaging Glass ampoule
Chemical Name or Material Cyclohexane-D12
Vapor Pressure 103 hPa (20°C)
Molecular Formula C6D12 (Hill)
CAS 1735-17-7
Ignition Point 260°C