Please login to your online account to display your discounted pricing

Cyclohexane-D12, deuteration degree min. 99.5%, for NMR spectroscopy, MagniSolv™, EMD Millipore™


For NMR Spectroscopy

Quantity 5mL
Packaging Glass ampoule

Manufacturer: emd millipore corporation  8150240006

 View more versions of this product

Catalog No. M8150240006

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 96.23
Quantity 5mL
Color Colorless
Physical Form Liquid
Chemical Name or Material Cyclohexane-D12
Vapor Pressure 103 hPa (20°C)
Molecular Formula C6D12 (Hill)
Odor Characterstic
Ignition Point 260°C
Assay Percent Range 99.5% min.
CAS 1735-17-7
Packaging Glass ampoule

Specifications

Chemical Name or Material Cyclohexane-D12
CAS 1735-17-7
Physical Form Liquid
Assay Percent Range 99.5% min.
Molecular Formula C6D12 (Hill)
Formula Weight 96.23