Please login to your online account to display your discounted pricing

Cyclohexane (Certified ACS), Fisher Chemical


Quantity 500mL
Packaging Glass Bottle

Manufacturer: fisher scientific company  BPC556500

 View more versions of this product

Catalog No. C556-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Boiling Point 81°C
Melting Point 6.5°C
Formula Weight 84.16
Quantity 500mL
Assay Percent Range ≥99.0%
DOT Information DOT Class 3, : Flammable Liquid
Flash Point -18°C (64.4°F)
Vapor Pressure 104mbar at 20°C
Purity Grade Certified ACS Reagent Grade
Viscosity 0.94 mPa/s at 20°C
Physical Form Liquid
Color Colorless
Molecular Formula C6H12
Density 0.770g/cm³
Identification Pass Test
Substances Darkened by H2SO4 Pass Test
Water 0.02% max.
Additional Information Vapor Pressure: 104mbar at 20°C
Packaging Glass Bottle
Residue after Evaporation 0.002% max.
Cleanup Symbol Solvent
MDL Number MFCD00003814
Synonym Hexahydrobenzene
Health Hazard 2 DANGER!
Health Hazard 3 Emergency Overview
Extremely flammable liquid and vapor. Inhalation may cause central nervous system effects. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:1
Flammability:3
Instability:0
Chemical Name or Material Cyclohexane
Linear Formula CH2(CH2)4CH2
CAS 110-82-7
ChemAlert Storage Symbol Red

Specifications

Chemical Name or Material Cyclohexane
CAS 110-82-7
Purity Grade Certified ACS Reagent Grade
Assay ≥99.0%
Assay Percent Range ≥99.0%
Molecular Formula C6H12
Linear Formula CH2(CH2)4CH2
Formula Weight 84.16
MDL Number MFCD00003814
Synonym Hexahydrobenzene
Accessories
 

Emergency Overview
Extremely flammable liquid and vapor. Inhalation may cause central nervous system effects. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:1
Flammability:3
Instability:0

DANGER!

 

SDS & Safety Documents