missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclohexane (Certified ACS), Fisher Chemical™

Supplier:  Thermo Fisher Scientific BPC5564

 View more versions of this product

Catalog No. C556-4




Description

Description

Specifications

Specifications

Cyclohexane
110-82-7
0.770g/cm³
4 L
Glass Bottle
0.002% max.
C6H12
MFCD00003814
0.94 mPa/s at 20°C
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
C1CCCCC1
84.16
CHEBI:29005
≥99.0%
6.5°C
Colorless
81°C
Pass Test
Liquid
≥99.0%
CH2(CH2)4CH2
104mbar at 20°C
0.02% max.
XDTMQSROBMDMFD-UHFFFAOYSA-N
cyclohexane
8078
84.16
Certified ACS Reagent

Chemical Identifiers

110-82-7
84.16
XDTMQSROBMDMFD-UHFFFAOYSA-N
8078
cyclohexane
C6H12
MFCD00003814
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
CHEBI:29005
C1CCCCC1
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Extremely flammable liquid and vapor. Inhalation may cause central nervous system effects. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:1
Flammability:3
Instability:0

DOTInformation : DOT Class 3, : Flammable Liquid

SDS Documents

Documents

Accessories
Promotions

Promotions