Please login to your online account to display your discounted pricing

Cyclohexane, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$124.43 - $1,337.15

Chemical Identifiers

Chemical Name or Material Cyclohexane
CAS 110-82-7
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula C6H12
Formula Weight 84.16
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002672

 
avantor performance
920622
Aluminum Bottle 1L Each for $279.86

02002673

 
avantor performance
920605
Aluminum Bottle 4L Each for $660.03

02002674

 
avantor performance
920633
Narrow Mouth PVC Coated Amber Bottle 4L Each for $1,337.15

02002677

 
avantor performance
920601
Narrow Mouth Amber Glass Bottle 500mL Each for $124.43
Description & Specifications

Specifications

Formula Weight 84.16
Purity Grade Baker Analyzed™ ACS Reagent
CAS 110-82-7
Specific Gravity 1L = 0.78kg
Chemical Name or Material Cyclohexane
Packaging Aluminum Bottle
Flash Point −18.00°C
Quantity 1L
Molecular Formula C6H12