missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclohexane, >99%, ACS, Alfa Aesar™

Maximum level of impurities: Color (APHA) 10 Evaporation residue 0.002%, Substances darkened by sulfuric acid P.T., H2O 0.02%

Manufacturer:  Alfa Aesar 22864K7

 View more versions of this product

Catalog No. AA22864K7



Specifications

Specifications

Cyclohexane
110-82-7
>99%
MFCD00003814
XDTMQSROBMDMFD-UHFFFAOYSA-N
cyclohexane
8078
84.16
ACS
245°C (473°F)
-18°C (64.4°F)
Colorless
4L
For ACS analysis
Colorless liquid
C6H12
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
C1CCCCC1
84.162
CHEBI:29005
Liquid
0.779g/cm3
Pungent
81°C
6.5°C

Chemical Identifiers

110-82-7
84.162
XDTMQSROBMDMFD-UHFFFAOYSA-N
8078
cyclohexane
C6H12
MFCD00003814
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
CHEBI:29005
C1CCCCC1
Safety and Handling

Safety and Handling

H225-H304-H400-H410-H315-H336: Highly flammable liquid and vapour. May be fatal if swallowed and enters airways. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Causes skin irritation. May cause drowsiness or dizziness.

DOTInformation : Hazard Class: 3, Packing Group: II

EINECSNumber : 203-806-2

SDS Documents