missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cyclohexane, 99.8%, for HPLC, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 279590025
Description
For HPLCSpecifications
Cyclohexane | |
110-82-7 | |
0.0002 meq/g max. | |
99.8% | |
C6H12 | |
05, 20 | |
benzene, hexahydro, benzenehexahydride, cicloesano, cyclohexaan, cyclohexan, cykloheksan, hexahydrobenzene, hexamethylene, hexanaphthene, polycyclohexane | |
C1CCCCC1 | |
84.162 | |
CHEBI:29005 | |
Crystalline Solid | |
HPLC | |
Yes | |
0.0002 % max. | |
White |
(1 cm cell vs water) | |
(1 cm cell vs water) | |
Vapor Pressure: 104mbar at 20°C | |
C6H12 | |
MFCD00003814 | |
15, 2716 | |
XDTMQSROBMDMFD-UHFFFAOYSA-N | |
cyclohexane | |
8078 | |
84.15 | |
≥99.8% (GC) | |
0.7700g/mL | |
Glass Bottle | |
0.01% Max. | |
2.5L |
Chemical Identifiers
110-82-7 | |
84.162 | |
XDTMQSROBMDMFD-UHFFFAOYSA-N | |
8078 | |
cyclohexane |
C6H12 | |
MFCD00003814 | |
benzene, hexahydro, benzenehexahydride, cicloesano, cyclohexaan, cyclohexan, cykloheksan, hexahydrobenzene, hexamethylene, hexanaphthene, polycyclohexane | |
CHEBI:29005 | |
C1CCCCC1 |
Safety and Handling
GHS P Statement May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Causes skin irritation. Very toxic to aquatic life with long lasting effects. Highly flammable liquid and vapor.
GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/ph
Danger
EINECSNumber : 203-806-2
RTECSNumber : GU6300000
TSCA : TSCA