Please login to your online account to display your discounted pricing

Alfa Aesar™ (Cyclohexadiene)iron tricarbonyl, 97+%

Chemical Identifiers

Chemical Name or Material (Cyclohexadiene)iron tricarbonyl
CAS 12152-72-6
Formula Weight 220.01
MDL Number MFCD00070435
UN Number UN3281
Synonym Tricarbonylcyclohexadiene iron
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3940302

  SDS
alfa aesar™
3940302
0.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3940304

  SDS
alfa aesar™
3940304
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 220.01
Health Hazard 1 Danger
Sensitivity Air Sensitive
Synonym Tricarbonylcyclohexadiene iron
Quantity 0.5g
MDL Number MFCD00070435
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, LIQUID, N.O.S.
Melting Point 8°C
UN Number UN3281
Assay ≥97%
Health Hazard 3 GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 12152-72-6
Health Hazard 2 GHS H Statement
H300-H330
Fatal if swallowed.
Fatal if inhaled.
Chemical Name or Material (Cyclohexadiene)iron tricarbonyl
Physical Form Liquid
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H330
Fatal if swallowed.
Fatal if inhaled.