missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cycloheptylamine 99%, ACROS Organics™

Manufacturer:  Acros Organics 111071000

 View more versions of this product

Catalog No. AC111071000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Cycloheptylamine
42°C
Glass bottle
0.89
54.0°C (11.0 mmHg)
100g
5452-35-7
C7H15N
MFCD00004153
cycloheptylamine, aminocycloheptane, cycloheptyl amine, cycloheptanamine, n-cycloheptylamine, 1-cycloheptylamine, cycloheptanyl amine, sz1, cycloheptylamine, acmc-209lh8
VXVVUHQULXCUPF-UHFFFAOYSA-N
cycloheptanamine
2899
Liquid
0.8900g/mL
Authentic
1.4710 to 1.4730
0.5% max. (K.F.)
Colorless to Yellow
99%
99%
C7H13NH2
12, 8
Solubility in water: soluble.
C1CCCC(CC1)N
113.204
113.2
99%

Chemical Identifiers

5452-35-7
113.204
VXVVUHQULXCUPF-UHFFFAOYSA-N
2899
C1CCCC(CC1)N
C7H15N
MFCD00004153
cycloheptylamine, aminocycloheptane, cycloheptyl amine, cycloheptanamine, n-cycloheptylamine, 1-cycloheptylamine, cycloheptanyl amine, sz1, cycloheptylamine, acmc-209lh8
cycloheptanamine
Safety and Handling

Safety and Handling

GHS P Statement
Flammable liquid and vapor.
Causes severe skin burns and eye damage.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Danger

EINECSNumber : 226-693-1

TSCA : TSCA

SDS Documents