missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cycloheptylamine 99%, ACROS Organics™

$76.01 - $76.01

Chemical Identifiers

CAS 5452-35-7
Molecular Formula C7H15N
Molecular Weight (g/mol) 113.204
MDL Number MFCD00004153
InChI Key VXVVUHQULXCUPF-UHFFFAOYSA-N
Synonym cycloheptylamine, aminocycloheptane, cycloheptyl amine, cycloheptanamine, n-cycloheptylamine, 1-cycloheptylamine, cycloheptanyl amine, sz1, cycloheptylamine, acmc-209lh8
PubChem CID 2899
IUPAC Name cycloheptanamine
SMILES C1CCCC(CC1)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC111070250 SDS View Documents Acros Organics
111070250
25g Glass bottle Each for $76.01
AC111071000 SDS View Documents Acros Organics
111071000
100g Glass bottle N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

5452-35-7
113.204
VXVVUHQULXCUPF-UHFFFAOYSA-N
2899
C1CCCC(CC1)N
C7H15N
MFCD00004153
cycloheptylamine, aminocycloheptane, cycloheptyl amine, cycloheptanamine, n-cycloheptylamine, 1-cycloheptylamine, cycloheptanyl amine, sz1, cycloheptylamine, acmc-209lh8
cycloheptanamine

Specifications

0.8900g/mL
Authentic
1.4710 to 1.4730
0.5% max. (K.F.)
Colorless to Yellow
5452-35-7
C7H15N
MFCD00004153
cycloheptylamine, aminocycloheptane, cycloheptyl amine, cycloheptanamine, n-cycloheptylamine, 1-cycloheptylamine, cycloheptanyl amine, sz1, cycloheptylamine, acmc-209lh8
VXVVUHQULXCUPF-UHFFFAOYSA-N
cycloheptanamine
2899
Liquid
Cycloheptylamine, 99%
42°C
Glass bottle
0.89
54.0°C (11.0 mmHg)
25g
99%
C7H13NH2
12, 8
Solubility in water: soluble.
C1CCCC(CC1)N
113.204
113.2
99%
Safety and Handling

Safety and Handling

GHS P Statement
Flammable liquid and vapor.
Causes severe skin burns and eye damage.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Danger

EINECSNumber : 226-693-1

TSCA : TSCA