Learn More
Cycloheptene, 96%, stab. with 0.1% BHT, Thermo Scientific Chemicals
$201.08 - $610.14
Chemical Identifiers
CAS | 628-92-2 |
---|---|
Molecular Formula | C7H12 |
Molecular Weight (g/mol) | 96.173 |
MDL Number | MFCD00004156 |
InChI Key | ZXIJMRYMVAMXQP-UHFFFAOYSA-N |
Synonym | cis-cycloheptene, z-cycloheptene, cyclohept-1-ene, 1-cycloheptene, z, trans-cyclohepten, e-cycloheptene, 1-cycloheptene #, cycloheptene, cycloheptene, pure |
PubChem CID | 12363 |
IUPAC Name | cycloheptene |
SMILES | C1CCC=CCC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH2906206
|
Thermo Scientific Chemicals
H2906206 |
5 g |
Each for $201.08
|
|
|||||
AAH2906214
|
Thermo Scientific Chemicals
H2906214 |
25 g |
Each for $610.14
|
|
|||||
Description
Cycloheptene was used in one-pot synthesis of 1,2/3-triols from the allylichydroperoxides catalyzed by zeolite-confined osmium(0) nanoclusters. It is a raw material in organic chemistry and a monomer in polymer synthesis. Copper (1) triflate catalyzes the photodimerization of the simple nonconjugated unstrained olefins, cyclopentene, cyclohexene, and cycloheptene, but not cyclooctene or acyclic olefins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCycloheptene was used in one-pot synthesis of 1,2/3-triols from the allylichydroperoxides catalyzed by zeolite-confined osmium(0) nanoclusters. It is a raw material in organic chemistry and a monomer in polymer synthesis. Copper (1) triflate catalyzes the photodimerization of the simple nonconjugated unstrained olefins, cyclopentene, cyclohexene, and cycloheptene, but not cyclooctene or acyclic olefins.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids. May react exothermically with reducing agents to release hydrogen gas.
Chemical Identifiers
628-92-2 | |
96.173 | |
ZXIJMRYMVAMXQP-UHFFFAOYSA-N | |
12363 | |
C1CCC=CCC1 |
C7H12 | |
MFCD00004156 | |
cis-cycloheptene, z-cycloheptene, cyclohept-1-ene, 1-cycloheptene, z, trans-cyclohepten, e-cycloheptene, 1-cycloheptene #, cycloheptene, cycloheptene, pure | |
cycloheptene |
Specifications
628-92-2 | |
112°C to 115°C | |
C7H12 | |
MFCD00004156 | |
UN2242 | |
Insoluble in water. | |
C1CCC=CCC1 | |
96.173 | |
96.17 | |
Cycloheptene, stabilized with 0.1% BHT |
0.833 | |
−7°C (19°F) | |
1.458 | |
5 g | |
cis-cycloheptene, z-cycloheptene, cyclohept-1-ene, 1-cycloheptene, z, trans-cyclohepten, e-cycloheptene, 1-cycloheptene #, cycloheptene, cycloheptene, pure | |
ZXIJMRYMVAMXQP-UHFFFAOYSA-N | |
cycloheptene | |
12363 | |
96% |
Safety and Handling
GHS H Statement
H225
Highly flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H225
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CYCLOHEPTENE
EINECSNumber : 211-060-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only