missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cycloheptanone, 98+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 111040250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Cycloheptanone | |
0.9500g/mL | |
55°C | |
98% min. (GC) | |
C7H12O | |
C7H12(=O) | |
25g | |
0.95 | |
suberone, ketocycloheptane, suberon, ketoheptamethylene, cycloheptanon, cycloheptyloxy, chembl18607, cycloheptan-1-one, cyclo-heptanone, 1-oxocycloheptane | |
CGZZMOTZOONQIA-UHFFFAOYSA-N | |
cycloheptanone | |
10400 | |
98+% |
502-42-1 | |
179.0°C | |
Authentic | |
Glass bottle | |
1.4600 to 1.4620 | |
MFCD00004159 | |
07, 13 | |
15, 2715 | |
Solubility in water: insoluble. Other solubilities: soluble in ether and alcohol | |
C1CCCC(=O)CC1 | |
112.17 | |
112.17 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
GHS Signal Word: Warning
EINECSNumber : 207-937-6
RUO â Research Use Only