Please login to your online account to display your discounted pricing

Cyclofenil, Tocris™

Selective estrogen receptor modulator (SERM)

$123.83 - $520.12

Chemical Identifiers

Chemical Name or Material Cyclofenil
Assay Percent Range >99%
Molecular Formula C23H24O4
Formula Weight 364.43
Synonym 4, 4'-(Cyclohexylidenemethylene)bisphenol 1, 1'-diacetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

399950

 
r & d systems
399950
50mg Each for $520.12

399910

 
r & d systems
399910
10mg Each for $123.83
Description & Specifications

Specifications

Formula Weight 364.43
Quantity 50mg
Chemical Name or Material Cyclofenil
Assay Percent Range >99%
Molecular Formula C23H24O4
CAS 2624-43-3
Synonym 4, 4'-(Cyclohexylidenemethylene)bisphenol 1, 1'-diacetate
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C