Learn More
Cyclododecyl isothiocyanate, 98%, Thermo Scientific™
$232.66 - $232.66
Chemical Identifiers
CAS | 59037-64-8 |
---|---|
Molecular Formula | C13H23NS |
Molecular Weight (g/mol) | 225.394 |
MDL Number | MFCD00041029 |
InChI Key | VSCLPFIUOOBLQJ-UHFFFAOYSA-N |
Synonym | cyclododecyl isothiocyanate, acmc-1atsj, cyclododecane,isothiocyanato, cyclododecylisothiocyanate |
PubChem CID | 143589 |
IUPAC Name | isothiocyanatocyclododecane |
SMILES | C1CCCCCC(CCCCC1)N=C=S |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1012006
|
Thermo Scientific Chemicals
L1012006 |
5 g |
Each for $232.66
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
59037-64-8 | |
225.394 | |
VSCLPFIUOOBLQJ-UHFFFAOYSA-N | |
143589 | |
C1CCCCCC(CCCCC1)N=C=S |
C13H23NS | |
MFCD00041029 | |
cyclododecyl isothiocyanate, acmc-1atsj, cyclododecane,isothiocyanato, cyclododecylisothiocyanate | |
isothiocyanatocyclododecane |
Specifications
59037-64-8 | |
C13H23NS | |
MFCD00041029 | |
UN1760 | |
Moisture sensitive | |
VSCLPFIUOOBLQJ-UHFFFAOYSA-N | |
isothiocyanatocyclododecane | |
143589 | |
98% |
0.99 | |
1.536 | |
5 g | |
4733327 | |
cyclododecyl isothiocyanate, acmc-1atsj, cyclododecane,isothiocyanato, cyclododecylisothiocyanate | |
C1CCCCCC(CCCCC1)N=C=S | |
225.394 | |
225.4 | |
Cyclododecyl isothiocyanate |
Safety and Handling
GHS H Statement
H301-H311-H314-H318-H332
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312+H332-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures