missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclododecene, 97%

Manufacturer:  Alfa Aesar™ 4470918

 View more versions of this product

Catalog No. AA4470918



Specifications

Specifications

Cyclododecene
93°C (199°F)
240°C
1501-82-2
MFCD00003721
HYPABJGVBDSCIT-OWOJBTEDSA-N
cyclododecene
5365627
Liquid
0.863g/mL
1.4882
50g
C12H22
e-cyclododecene, cyclododecene, e, trans-cyclododecene, 1e-cyclododecene, t-cyclododecene, cyclododecene, trans, 11e-cyclododecene
C1CCCCCC=CCCCC1
166.308
160.31
97%

Chemical Identifiers

1501-82-2
166.308
HYPABJGVBDSCIT-OWOJBTEDSA-N
5365627
C1CCCCCC=CCCCC1
C12H22
MFCD00003721
e-cyclododecene, cyclododecene, e, trans-cyclododecene, 1e-cyclododecene, t-cyclododecene, cyclododecene, trans, 11e-cyclododecene
cyclododecene
SDS Documents