Please login to your online account to display your discounted pricing

Alfa Aesar™ Cyclododecene, 97%

$52.74 - $203.42

Chemical Identifiers

Chemical Name or Material Cyclododecene
CAS 1501-82-2
Molecular Formula C12H22
Formula Weight 160.31
MDL Number MFCD00003721
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4470918

  SDS
alfa aesar™
4470918
50g Each for $52.74

AA4470930

  SDS
alfa aesar™
4470930
250g Each for $203.42
Description & Specifications

Specifications

Formula Weight 160.31
Chemical Name or Material Cyclododecene
Boiling Point 240°C
Molecular Formula C12H22
Physical Form Liquid
MDL Number MFCD00003721
Refractive Index 1.4882
Quantity 50g
Density 0.863g/mL
Flash Point 93°C (199°F)
CAS 1501-82-2
Assay 97%