missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cyclododecanone, TCI America™

$22.92 - $128.34

Chemical Identifiers

CAS 830-13-7
Molecular Formula C12H22O
Molecular Weight (g/mol) 182.307
MDL Number MFCD00003722
InChI Key SXVPOSFURRDKBO-UHFFFAOYSA-N
Synonym cyclododecan-1-one, dsstox_cid_7322, dsstox_rid_78407, dsstox_gsid_27322, cyclododecanon, cdon, cyclododecanone, acmc-209pr1, ksc448s4j
PubChem CID 13246
IUPAC Name cyclododecanone
SMILES C1CCCCCC(=O)CCCCC1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C046225G View Documents TCI America
C046225G
25g Each for $22.92
 
C0462500G View Documents TCI America
C0462500G
500g Each for $128.34
 
C0462100G View Documents TCI America
C0462100G
100g Each for $42.78
 
Specifications

Chemical Identifiers

830-13-7
182.307
SXVPOSFURRDKBO-UHFFFAOYSA-N
13246
C1CCCCCC(=O)CCCCC1
C12H22O
MFCD00003722
cyclododecan-1-one, dsstox_cid_7322, dsstox_rid_78407, dsstox_gsid_27322, cyclododecanon, cdon, cyclododecanone, acmc-209pr1, ksc448s4j
cyclododecanone

Specifications

113°C
62°C
830-13-7
MFCD00003722
cyclododecan-1-one, dsstox_cid_7322, dsstox_rid_78407, dsstox_gsid_27322, cyclododecanon, cdon, cyclododecanone, acmc-209pr1, ksc448s4j
C1CCCCCC(=O)CCCCC1
182.307
182.31
≥99.0% (GC)
White
25g
C12H22O
3077
SXVPOSFURRDKBO-UHFFFAOYSA-N
cyclododecanone
13246
Crystal-Powder at 20°C
Cyclododecanone
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2379

TSCA : Yes