Please login to your online account to display your discounted pricing

Cyclododecanone, 99+%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: fisher scientific acros  110975000

 View more versions of this product

Catalog No. AC110975000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Adhering Crystals
Assay ≥99.0%
Formula Weight 182.31
Quantity 500g
Color Colorless to white
Beilstein 07, II, 48
EINECS Number 212-595-6
TSCA TSCA
Melting Point 59° to 61°C
Chemical Name or Material Cyclododecanone
Name Note 99+%
Assay Percent Range 99+%
Packaging Glass bottle
Molecular Formula C12H22O
Linear Formula C12H122(=O)
MDL Number MFCD00003722
CAS 830-13-7
Health Hazard 2 GHS H Statement:
Toxic to aquatic life with long lasting effects.
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Cyclododecanone
Name Note 99+%
CAS 830-13-7
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C12H22O
Linear Formula C12H122(=O)
Formula Weight 182.31
MDL Number MFCD00003722
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic to aquatic life with long lasting effects.

SDS & Safety Documents