Please login to your online account to display your discounted pricing

Cyclododecanone, 99+%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  110971000

 View more versions of this product

Catalog No. AC110971000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Adhering Crystals
Assay ≥99.0%
Formula Weight 182.31
Quantity 100g
Color Colorless to white
Beilstein 07, II, 48
TSCA TSCA
Melting Point 59° to 61°C
EINECS Number 212-595-6
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Toxic to aquatic life with long lasting effects.
MDL Number MFCD00003722
Name Note 99+%
Assay Percent Range 99+%
Linear Formula C12H122(=O)
CAS 830-13-7
Chemical Name or Material Cyclododecanone
Molecular Formula C12H22O
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Cyclododecanone
Name Note 99+%
CAS 830-13-7
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C12H22O
Linear Formula C12H122(=O)
Formula Weight 182.31
MDL Number MFCD00003722
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic to aquatic life with long lasting effects.

SDS & Safety Documents